Läs mer
Acetyl-Alpha-boswellic Acid, TRC
Produktkod. 30326856
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30326856 10 mg
1 options
Produktkod. 30326856 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA14513510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Acetyl-alpha-Boswellic acid
CAS 89913-60-0
Molekylformel C32 H50 O4
Kvantitet 10 mg
Synonym Olean-12-en-23-oic acid, 3-(acetyloxy)-, (3α,4β)-, (3α,4β)-3-(Acetyloxy)olean-12-en-23-oic acid, 3-O-Acetyl-α-boswellic acid, Acetyl-α-boswellic acid, α-Boswellic acid acetate
InChI formel InChI=1S/C32H50O4/c1-20(33)36-25-12-13-29(5)23-10-9-21-22-19-27(2,3)15-16-28(22,4)17-18-30(21,6)31(23,7)14-11-24(29)32(25,8)26(34)35/h9,22-25H,10-19H2,1-8H3,(H,34,35)/t22-,23+,24+,25+,28+,29+,30+,31+,32+/m0/s1
LEDER CC(=O)O[C@@H]1CC[C@@]2(C)[C@@H](CC[C@]3(C)[C@@H]2CC=C4[C@@H]5CC(C)(C)CC[C@]5(C)CC[C@@]34C)[C@@]1(C)C(=O)O
IUPAC-namn (3R,4R,4aR,6aR,6bS,8aR,12aR,14aR,14bR)-3-acetyloxy-4,6a,6b,8a,11,11,14b-heptamethyl-1,2,3,4a,5,6,7,8,9,10,12,12a,14,14a-tetradecahydropicene-4-carboxylic acid
Molekylvikt (g/mol) 498.74
Formel vikt 498.3709