Läs mer
Acetyl-D-alpha-aminobutyric acid, TRC
Produktkod. 30318353
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30318353 25 mg
1 options
Produktkod. 30318353 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA29812525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Acetyl-D-alpha-aminobutyric acid
CAS 34271-27-7
Molekylformel C6H11NO3
Kvantitet 25 mg
Synonym (R)-2-Acetamidobutanoic Acid
InChI formel InChI=1S/C6H11NO3/c1-3-5(6(9)10)7-4(2)8/h5H,3H2,1-2H3,(H,7,8)(H,9,10)/t5-/m1/s1
LEDER CC[C@@H](NC(=O)C)C(=O)O
IUPAC-namn (2R)-2-acetamidobutanoic acid
Molekylvikt (g/mol) 145.16
Formel vikt 145.0739