Läs mer
Acetyl Propionate, TRC
Produktkod. 30324499
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30324499 100 mg
1 options
Produktkod. 30324499 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA167668100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Acetic Propionic Anhydride
CAS 13080-96-1
Molekylformel C5 H8 O3
Kvantitet 100 mg
Synonym Propanoic acid, anhydride with acetic acid, Propionic acid, anhydride with AcOH (6CI,7CI), Propionic acid, anhydride with acetic acid (8CI), Acetic acid, anhydride with propionic acid (8CI), Acetic propionic anhydride, Acetyl propionate
InChI formel InChI=1S/C5H8O3/c1-3-5(7)8-4(2)6/h3H2,1-2H3
LEDER CCC(=O)OC(=O)C
IUPAC-namn acetyl propanoate
Molekylvikt (g/mol) 116.1152
Formel vikt 116.0473