Läs mer
Acetylcedrene (~80%), TRC
Produktkod. 30010042
Change view
Kvantitet:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010042 10 g
30012624 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010042 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA17198510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 32388-55-9
Molekylformel C17 H26 O
Molekylvikt (g/mol) 246.39
Synonym alpha-Acetylcedrene, Ethanone, 1-(2,3,4,7,8,8a-hexahydro-3,6,8,8-tetramethyl-1H-3a,7-methanoazulen-5-yl)-, [3R-(3α,3aβ,7β,8aα)]-, 1-[(3R,3aR,7R,8aS)-2,3,4,7,8,8a-Hexahydro-3,6,8,8-tetramethyl-1H-3a,7-methanoazulen-5-yl]ethanone, 1H-3a,7-Methanoazulene, ethanone deriv., 1-((3R,3AR,7R,8aS)-3,6,8,8-tetramethyl-2,3,4,7,8,8a-hexahydro-1H-3a,7-methanoazulen-5-yl)ethan-1-one, 1-[(3R,3aR,7R,8aS)-3,6,8,8-Tetramethyl-2,3,4,7,8,8a-hexahydro-1H-3a,7-methanoazulen-5-yl]ethan-1-one, 1-[(3R,3aR,7R,8aS)-3,6,8,8-Tetramethyl-2,3,4,7,8,8a-hexahydro-1H-3a,7-methanoazulen-5-yl]ethan-1-one, Acetyl-α-cedrene, Acetylcedrene, Lixetone, Vertofix, Ethanone, 1-[(3R,3aR,7R,8aS)-2,3,4,7,8,8a-hexahydro-3,6,8,8-tetramethyl-1H-3a,7-methanoazulen-5-yl]-, Acetyl-alpha-cedrene, 1-[(3R,3aR,7R,8aS)-2,3,4,7,8,8a-Hexahydro-3,6,8,8-tetramethyl-1H-3a,7-methanoazulen-5-yl]ethanone, 1H-3a,7-Methanoazulene, ethanone deriv., 1-[(3R,3aR,7R,8aS)-3,6,8,8-Tetramethyl-2,3,4,7,8,8a-hexahydro-1H-3a,7-methanoazulen-5-yl]ethan-1-one, 1-[(3R,3aR,7R,8aS)-3,6,8,8-Tetramethyl-2,3,4,7,8,8a-hexahydro-1H-3a,7-methanoazulen-5-yl]ethan-1-one, Acetyl-α-cedrene, Acetylcedrene, Lixetone, Vertofix
IUPAC-namn 1-[(1R,2R,5S,7R)-2,6,6,8-tetramethyl-9-tricyclo[5.3.1.01,5]undec-8-enyl]ethanone
LEDER C[C@@H]1CC[C@H]2C(C)(C)[C@H]3C[C@@]12CC(=C3C)C(=O)C
Specifikationer
Kemiskt namn eller material Acetyl-alpha-cedrene
Analyt- eller komponentnamn Acetyl-alpha-cedrene
Frakt skick Room Temperature
InChI formel InChI=1S/C17H26O/c1-10-6-7-15-16(4,5)14-9-17(10,15)8-13(11(14)2)12(3)18/h10,14-15H,6-9H2,1-5H3/t10-,14+,15+,17+/m1/s1
Lösningstyp Neat
Industry Type Forensics
Kvantitet 10 g
Formel vikt 246.1984