Läs mer
Acetylhistamine, TRC
Produktkod. 30312321
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30312321 5 mg
30326147 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30312321 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1791105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Acetylhistamine
CAS 673-49-4
Molekylformel C7H11N3O
Kvantitet 5 mg
Synonym N-[2-(1H-Imidazol-5-yl)ethyl]acetamide, N-[2-imidazol-4(or 5)-ylethyl]acetamide
InChI formel InChI=1S/C7H11N3O/c1-6(11)9-3-2-7-4-8-5-10-7/h4-5H,2-3H2,1H3,(H,8,10)(H,9,11)
LEDER CC(NCCC1=CN=CN1)=O
IUPAC-namn N-[2-(1H-imidazol-5-yl)ethyl]acetamide
Molekylvikt (g/mol) 153.18
Formel vikt 153.0902