Läs mer
Acetylpyruvic Acid, TRC
Produktkod. 30317464
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30317464 1 g
1 options
Produktkod. 30317464 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1639401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Acetylpyruvic Acid
CAS 5699-58-1
Molekylformel C5H6O4
Kvantitet 1 g
Synonym 2,4-Dioxopentanoic Acid, Acetoneoxalic Acid, Acetopyruvic Acid, Acetylpyruvic Acid, 2,4-Dioxo-valeric Acid, 2,4-Dioxo-pentanoic Acid
InChI formel InChI=1S/C5H6O4/c1-3(6)2-4(7)5(8)9/h2H2,1H3,(H,8,9)
LEDER CC(=O)CC(=O)C(=O)O
IUPAC-namn 2,4-dioxopentanoic acid
Molekylvikt (g/mol) 130.1
Formel vikt 130.0266