Läs mer
Acetylsalicylic Acid Acyl-b-D-glucuronide (>85%), TRC
Produktkod. 30132558
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132558 10 mg
30004107 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132558 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA18777510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 24719-72-0
Molekylformel C15 H16 O10
Molekylvikt (g/mol) 356.28
Synonym β-D-Glucopyranuronic acid, 1-[2-(acetyloxy)benzoate], Glucopyranuronic acid, 1-salicylate monoacetate, β-D- (8CI), Salicylic acid acetate, 1-ester with β-D-glucopyranosiduronic acid (8CI)
IUPAC-namn (2S,3S,4S,5R,6S)-6-(2-acetyloxybenzoyl)oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
LEDER CC(=O)Oc1ccccc1C(=O)O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O
Specifikationer
Kemiskt namn eller material Acetylsalicylic Acid Acyl-beta-D-glucuronide
Kvantitet 10 mg
InChI formel InChI=1S/C15H16O10/c1-6(16)23-8-5-3-2-4-7(8)14(22)25-15-11(19)9(17)10(18)12(24-15)13(20)21/h2-5,9-12,15,17-19H,1H3,(H,20,21)/t9-,10-,11+,12-,15-/m0/s1
Formel vikt 356.0743