Läs mer
Acetylsalicylic Acid EP Impurity B (Salicylic Acid USP Related Compound B), Mikromol™
Beskrivning
Acetylsalicylic Acid EP Impurity B (Salicylic Acid USP Related Compound B)
Specifikationer
Specifikationer
| Kemiskt namn eller material | 4-Hydroxyisophthalic Acid |
| CAS | 636-46-4 |
| Analyt- eller komponentnamn | 4-Hydroxyisophthalic Acid |
| Frakt skick | Room Temperature |
| Molekylformel | C8 H6 O5 |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | Acetylsalicylic Acid Imp. B (EP), Salicylic Acid Imp. B (EP), Salicylic Acid USP Related Compound B, Salicylic Acid USP RC B, 4-Hydroxybenzene-1,3-dicarboxylic Acid, 4-Hydroxyisophthalic Acid, 1,3-Benzenedicarboxylic acid, 4-hydroxy-, Isophthalic acid, 4-hydroxy- (6CI,7CI,8CI), 4-Hydroxy-1,3-benzenedicarboxylic acid, 4-Hydroxy-m-phthalic acid, Eupirina, NSC 2445, Ph Eur Salicylic Acid Impurity B, USP Salicylic Acid Related Compound B, Salicylic Acid Related Compound B, Acetylsalicylic Acid Impurity B, Salicylic Acid Impurity B |
| InChI formel | InChI=1S/C8H6O5/c9-6-2-1-4(7(10)11)3-5(6)8(12)13/h1-3,9H,(H,10,11)(H,12,13) |
| LEDER | OC(=O)c1ccc(O)c(c1)C(=O)O |
| IUPAC-namn | 4-hydroxybenzene-1,3-dicarboxylic acid |
| Visa mer |