missing translation for 'onlineSavingsMsg'
Läs mer

Acetylsalicylsalicylsalicylic Acid, Mikromol™

Produktkod. 30050605
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050605 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30050605 Leverantör Mikromol Leverantörsnummer MM0133.090025

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Acetylsalicylsalicylsalicylic Acid
TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Acetylsalicylsalicylsalicylic Acid
CAS 85531-16-4
Analyt- eller komponentnamn Acetylsalicylsalicylsalicylic Acid
Frakt skick Room Temperature
Molekylformel C23 H16 O8
Certifikat och efterlevnad ISO 17025
Synonym Acetylsalicylsalicylsalicylic Acid
InChI formel InChI=1S/C23H16O8/c1-14(24)29-19-12-6-3-9-16(19)22(27)31-20-13-7-4-10-17(20)23(28)30-18-11-5-2-8-15(18)21(25)26/h2-13H,1H3,(H,25,26)
LEDER CC(=O)Oc1ccccc1C(=O)Oc2ccccc2C(=O)Oc3ccccc3C(=O)O
IUPAC-namn 2-[2-(2-acetyloxybenzoyl)oxybenzoyl]oxybenzoic acid
Molekylvikt (g/mol) 420.37
Kvantitet 25 mg
Formel vikt 420.0845 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.