Description
- Detection method: Fluorescent
- Applications: Enzyme activity assay
- Sample types: Purified protein, Inhibitor compounds
- Equipment: Microplate reader
- Citation count: 5
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Detection Location | In-Solution Detection |
| For Use With (Equipment) | Microplate Reader |
| Format | Kit |
| No. of Reactions | 96 Tests |
| Target Molecule | Acetyltransferase (ACAT2) |
| Detection Method | Fluorescence |
| Sample Type | Purified protein, Inhibitor compounds |
| Quantity | 96 Tests |
| For Use With (Application) | Enzyme activity assay |