missing translation for 'onlineSavingsMsg'
Läs mer

Acid Blue 113 (Technical Grade), TRC

Produktkod. 30008443
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008443 10 mg
30004809 50 mg
30005991 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30008443 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA18986310MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 3351-05-1
Molekylformel C32H21N5Na2O6S2
Molekylvikt (g/mol) 681.65
Synonym 1-Naphthalenesulfonic acid, 8-(phenylamino)-5-[[4-[(3-sulfophenyl)azo]-1-naphthalenyl]azo]-, disodium salt (9CI), C.I. Acid Blue 113 (7CI), C.I. Acid Blue 113, disodium salt (8CI), Acid Blue 5R, Acid Blue S 5R, Acid Blue for Leather, Acid Cyanine 5R, Acid Cyanine Blue 5R, Acid Dark Blue 2G, Acid Dark Blue for Polyamide, Acid Fast Blue 5R, Acid Fast Blue N 5R, Acid Leather Blue 5RN, Acid Leather Blue RN, Acid Milling Navy Blue 2RNX, Acid Milling Navy Blue S 5R, Acid Navy Blue 2BN, Acid Navy Blue 2RHX, Acid Navy Blue 2RNX, Acid Navy Blue 5R, Alphanol Blue 5RN, Amacid Fast Navy 5R, Ambinyl Cyanine M 5R, Anadurm Navy M-5RX, Apollo Acid Cyanine 5R, Atul Acid Cyanine Blue 5R, Azo Cyanine Blue 5R Extra, Benzyl Fast Blue 5R, Best Acid Milling Cyanine 5R, C.I. 26360, Cirene Blue 5RA, Cloth Blue 5R, Cloth Fast Blue R, Cloth Fast Blue RN, Cloth Fast Blue S 5R, Colacid Fast Blue 5R, Colocid Fast Navy Blue 5R, Colorosacid Blue 113, Conacid Blue AAC, Concorde Acid Cyanine M 5R, Concorde Leather Blue 5R, Coomassie Navy Blue 2RN, Covalene Navy 2R, Covanyl Navy 2R, Cyanine 5R, Cyanine Navy Blue 2RNX, Cyanine Navy Blue 5R, Daedo Acid Navy Blue BNC, Dermacid Blue LN, Dinacid Navy Blue 2RNS, Dycosweak Acid Navy Blue 5R, Eniacid Blue 5R, Eniacid Cloth Blue 5R, Erio Fast Blue S 5R, Erionyl Blue S 5R, Erionyl Navy A-R, Erionyl Navy Blue 2RN, Erionyl Navy R, Eriosin Fast Blue S 5R, Everacid Cyanine N 5R, Everacid Milling Cyanine 5R, Everacid Navy 2RN, Fabracid Navy Blue M-SR, Fast Blue 5R, Fenazo Blue S 5R, Folan Fast Navy Blue 5R, Follone Blue 5R, Formo Blue 5R, Hispacid Cloth Blue 5RF, Indacid Milling Navy Blue S 5R, Intrazone Fast Blue 5RS, Java Fast Blue 5R, Kayanol Cyanine 5R, Kayanol Milling Cyanine 5R, Kemacid Blue 2RNX, Kenamide Navy K 2R, Kenanthrol Navy 2R, Leather Cadet Blue RB, Lecotan Navy Blue 2R, Levanol Fast Cyanine 5RN, Libacid Navy Blue L 2RNX, Midlon Blue 5RG, Milling Fast Blue 5R, Milling Navy Blue 2RNX, Monacid Navy Blue 2RNX, Multicuer Blue 5R, Nanhacid Navy N-RBL, Naphthalene Leather Navy 2RN, Navitan Fast Blue S 5R, Navy Blue 5R, Navy N-BL, Navy N-RBL, Neutral Blue 5R, Neutral Fast Blue 5R, Nylomine Acid Navy C 2R, Nylomine Acid Navy P 2R, Nylosan Navy N-BL, Nylosan Navy N-RBL, Nylosan Navy N-RBLA, Nylosan Navy N-RE, Optanol Cyanine 5R extra, Pacid Navy Blue 2RNX, Polan Navy Blue R, Pontacyl Fast Blue 5R, Ranocid Fast Blue S 5R, Rifa Acid Cyanine 5R, Rifa Acid Navy Blue BN, Rifa Leather Navy Blue 2RN, Rybanyl Navy NL 5R, Sella Fast Navy C-R, Simacid Navy Blue 24914, Sulfocyanine Blue 5R, Sulfon Blue 5R, Sulfone Cyanin 5R, Sulfonine Blue 5R, Sulphon Cyanine 5RA-CF, Sulphoncin Blue 5R, Sulphoncyanine 5R Extra, Sulphonol Navy 5RX, Suminol Cyanine 5R extra, Suminol Milling Cyanine 5R, Suminol Milling Cyanine 5R Extra, Supranol Fast Cyanin 5R, Supranol Fast Cyanine 5R, Supranol Fast Cyanine 5R Extra, Supranol Fast Cyanine 5RT Extra, Supranol Navy R, Telon Fast Navy Blue R, Telon Navy AMF, Telon Supra Navy Blue RN, Tertracid Fast Navy Blue CR, Tracid Cyanine 5R, Triacid Milling Navy Blue 5RN, Triacid Sulfon Blue 5R, Vicoacid Blue 113, Victacid Blue 5R, Vondamol Fast Blue 5R, Vopsider Blue A 5RL, Vopsider Blue N-A 5RL, Wool Fast Blue 5R, Wool Fast Navy Blue 5R, Yamada Acid Cyanine 5R
IUPAC-namn disodium;8-anilino-5-[(E)-[4-[(E)-(3-sulfonatophenyl)diazenyl]naphthalen-1-yl]diazenyl]naphthalene-1-sulfonate
LEDER O=S(C1=CC=CC(/N=N/C2=C3C=CC=CC3=C(/N=N/C4=C(C=CC=C5S(=O)(O[Na])=O)C5=C(NC6=CC=CC=C6)C=C4)C=C2)=C1)(O[Na])=O

Specifikationer

Kemiskt namn eller material Acid Blue 113
Kvantitet 10 mg
InChI formel InChI=1S/C32H23N5O6S2.2Na/c38-44(39,40)23-11-6-10-22(20-23)34-35-27-16-17-28(25-13-5-4-12-24(25)27)36-37-29-18-19-30(33-21-8-2-1-3-9-21)32-26(29)14-7-15-31(32)45(41,42)43;;/h1-20,33H,(H,38,39,40)(H,41,42,43);;/q;2*+1/p-2
Formel vikt 681.0729
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.