missing translation for 'onlineSavingsMsg'
Läs mer

Acid Green 50 (Technical Grade), TRC

Produktkod. 30323219
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30323219 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323219 Supplier Toronto Research Chemicals Supplier No. TRCA18989510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Acid Green 50
CAS 3087-16-9
Molekylformel C27 H25 N2 O7 S2 . Na
Kvantitet 10 g
Synonym C.I. Acid Green 50 (7CI), C.I. Acid Green 50, monosodium salt (8CI), Lissamine Green BN (6CI), Methanaminium, N-[4-[[4-(dimethylamino)phenyl](2-hydroxy-3,6-disulfo-1-naphthalenyl)methylene]-2,5-cyclohexadien-1-ylidene]-N-methyl-, hydroxide, inner salt, monosodium salt, Methanaminium, N-[4-[[4-(dimethylamino)phenyl](2-hydroxy-3,6-disulfo-1-naphthalenyl)methylene]-2,5-cyclohexadien-1-ylidene]-N-methyl-, inner salt, monosodium salt (9CI), 12078 Green, Acid Brilliant Green BS, Acid Green 50, Acid Green BS, Acid Green RS, Acid Leather Green S, Acidal Wool Green BS, Acilan Green BS, Amacid Wool Green S, Ariavit Green S, Brilliant Acid Green BS, Bucacid Wool Green, C.I. 44090, C.I. Food Green 4, Calcocid Green S, Calcocid Green SB, Dycosacid Green BS, E 142, Edicol Supra Green B, Edicol Supra Green BS, Erio Green S, Eurocert Green S 312524, Green H 002, Green S, Green S 44090, Hexacol Green S, Hidacid Wool Green, Kiton Green S, Lissamine Green B, Naphthazine Green S, Pharmacid Green S, Sumitomo Wool Green S, Unitertracid Green BS, Vondacid Green S, Water Green 176558, Water Green SX, Wool Green BS, Wool Green BSNA, Wool Green MS, Wool Green S, Wool Green SF, Wool Green SG
InChI formel InChI=1S/C27H26N2O7S2.Na/c1-28(2)20-9-5-17(6-10-20)25(18-7-11-21(12-8-18)29(3)4)26-23-14-13-22(37(31,32)33)15-19(23)16-24(27(26)30)38(34,35)36;/h5-16H,1-4H3,(H2,31,32,33,34,35,36);/q;+1/p-1
LEDER [Na+].CN(C)c1ccc(cc1)C(=C2C=CC(=[N+](C)C)C=C2)c3c(O)c(cc4cc(ccc34)S(=O)(=O)[O-])S(=O)(=O)[O-]
IUPAC-namn sodium;4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]-3-hydroxynaphthalene-2,7-disulfonate
Molekylvikt (g/mol) 576.62
Formel vikt 576.1001
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.