Läs mer
Acid Orange 24, Technical Grade, TRC
Produktkod. 30136685
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136685 25 mg
30134085 10 mg
18031640 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136685 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA18983525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1320-07-6
Molekylformel C20 H17 N4 O5 S . Na
Molekylvikt (g/mol) 448.43
Synonym Benzenesulfonic acid, 4-[[3-[(dimethylphenyl)azo]-2,4-dihydroxyphenyl]azo]-, monosodium salt (9CI), C.I. Acid Orange 24 (7CI), C.I. Acid Orange 24, monosodium salt (8CI), Acid Orange 24, Albion Leather Brown FH, Brown 201, Brown No. 201, C Ext. Brown 4, C.I. 20170, D & C Brown 1, D and C Brown No. 1, D&C Brown No. 1, Fabracid Brown S-B, Japan Brown 201, Japan Brown No. 201, Leather Brown 2G, Leather Brown RGN, Multicuer Brown MNO, Ratna Acid Fast Brown 2BB, Resorcin Brown, Resorcin Brown G, Resorcin Brown R, Resorcine Brown B, Resorcine Brown G, Resorcine Brown R;Sodium 4-((2,4-Dimethylphenyl)diazenyl)-2,4-dihydroxyphenyl)diazenyl)benzenesulfonate, Sodium,4-[(2Z)-2-[(5E)-5-[(2,4-dimethylphenyl)hydrazinylidene]-4,6-dioxocyclohex-2-en-1-ylidene]hydrazinyl]benzenesulfonate, C.I. Acid Orange 24, Monosodium Salt, Albion Leather Brown FH, Brown 201, Brown No. 201, C Ext. Brown 4, C.I. 20170, D & C Brown 1, D and C Brown No. 1, D&C Brown No. 1, Fabracid Brown S-B, Japan Brown 201, Japan Brown No. 201, Leather Brown 2G, Leather Brown RGN, Multicuer Brown MNO, Ratna Acid Fast Brown 2BB, Resorcin Brown, Resorcin Brown G, Resorcin Brown R, Resorcine Brown B, Resorcine Brown G, Resorcine Brown R
IUPAC-namn sodium;4-[(E)-[3-[(Z)-(2,4-dimethylphenyl)azo]-2,4-dihydroxy-phenyl]azo]benzenesulfonate
LEDER [Na+].Cc1ccc(N=Nc2c(O)ccc(N=Nc3ccc(cc3)S(=O)(=O)[O-])c2O)c(C)c1
Specifikationer
Kemiskt namn eller material Acid Orange 24
Analyt- eller komponentnamn Acid Orange 24
Frakt skick Room Temperature
InChI formel InChI=1S/C20H18N4O5S.Na/c1-12-3-8-16(13(2)11-12)22-24-19-18(25)10-9-17(20(19)26)23-21-14-4-6-15(7-5-14)30(27,28)29;/h3-11,25-26H,1-2H3,(H,27,28,29);/q;+1/p-1/b23-21+,24-22-;
Lösningstyp Neat
Industry Type Forensics
Kvantitet 25 mg
Formel vikt 448.0817