Läs mer
Acid Orange 8 (Technical Grade), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Acid Orange 8 (Technical Grade) |
| CAS | 5850-86-2 |
| Molekylformel | C17H13N2NaO4S |
| Kvantitet | 2.5 g |
| Synonym | 4-[(2-Hydroxy-1-naphthalenyl)azo]-3-methylbenzenesulfonic Acid Monosodium Salt, C.I. Acid Orange 8, monosodium salt (8CI)Acid Brilliant Orange RAcid Orange 8Acid Orange ROAiredale Orange RBasacid Orange 288Brilliant Orange RBrilliant Orange ROC.I. 15575C.I. Acid Orange 8Calcocid Orange RRDiacid Orange RKayaku Acid Orange ROKeyacid Orange ROKiton Fast Orange 2RKiton Orange RNaphthazine Orange ROrange 2ROOrange ROrange ROOrange TPolycor Orange RRitacid Orange ROShikiso Acid Orange ROSolar Orange RTertracid Orange ROTriacid Orange ROVondacid Orange ROWool Orange RYodogawa Acid Orange R |
| InChI formel | InChI=1S/C17H14N2O4S.Na/c1-11-10-13(24(21,22)23)7-8-15(11)18-19-17-14-5-3-2-4-12(14)6-9-16(17)20;/h2-10,20H,1H3,(H,21,22,23);/q;+1/p-1 |
| LEDER | [Na+].Cc1cc(ccc1N=Nc2c(O)ccc3ccccc23)S(=O)(=O)[O-] |
| IUPAC-namn | sodium;4-[(E)-(2-hydroxynaphthalen-1-yl)diazenyl]-3-methylbenzenesulfonate |
| Molekylvikt (g/mol) | 364.35 |
| Formel vikt | 364.0494 |