Läs mer
Acid Red 249, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Acid Red 249 |
| CAS | 6416-66-6 |
| Molekylformel | C29 H20 Cl N3 Na2 O10 S3 |
| Kvantitet | 50 mg |
| Synonym | Acid Brilliant Red B, Acid Red 249, Anadurm Brilliant Red M-B, Benzyl Fast Rubine 2B, Best Acid Milling Red 2B, Best Acid Red 2B, Brilliant Milling Red 2B, Brilliant Milling Red B, Brilliant Red B, C.I. 18134, C.I. Acid Red 249, Chuganol Milling Brilliant Red B Conc, Colomill Red 2BN, Concorde Acid Red 2B, Dycosweak Acid Brilliant, Red B, Dynacid Red BM, Elbenyl Red C 2B, Elite Fast Brilliant Red B, Euroderm Red 2BL, Everacid Red B, Everacid Red N-B, Kenanthrol Red BN, Milling Brilliant Red M 2BN, Naphthazine Milling Brilliant Red BN, Polar Brilliant Red B, Polar Red B, Pro-Jet Magenta 1, Pro-Jet Magneta 1, Pula Red B, Sandolan Brilliant Red N-BG, Sandolan Brilliant Red N-BG 125, Sulfonine Brilliant Red B, Sulfonine Brilliant Red BG, Suminol Milling Brilliant Red B, Supranol Brilliant Red BB, Synoacid Red F-B, Tertracid Brilliant Milling Red, Tertracid Brilliant Milling Red 2B, Tracid Brilliant Red B, Triacid Fast Red 2BN, Vicoacid Red 249, Weak Acid Brilliant Red B |
| InChI formel | InChI=1S/C29H22ClN3O10S3.2Na/c1-17-7-10-21(11-8-17)44(35,36)33-24-16-22(45(37,38)39)13-18-14-26(46(40,41)42)28(29(34)27(18)24)32-31-23-15-19(30)9-12-25(23)43-20-5-3-2-4-6-20;;/h2-16,33-34H,1H3,(H,37,38,39)(H,40,41,42);;/q;2*+1/p-2/b32-31+;; |
| LEDER | Cc1ccc(cc1)S(=O)(=O)Nc2cc(cc3cc(c(N=Nc4cc(Cl)ccc4Oc5ccccc5)c(O)c23)S(=O)(=O)O[Na])S(=O)(=O)O[Na] |
| IUPAC-namn | [3-[(E)-(5-chloro-2-phenoxy-phenyl)azo]-4-hydroxy-5-(p-tolylsulfonylamino)-7-sodiooxysulfonyl-2-naphthyl]sulfonyloxysodium |
| Molekylvikt (g/mol) | 748.11 |
| Formel vikt | 746.9795 |