Läs mer
Acid Red 35, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Acid Red 35 |
| CAS | 6441-93-6 |
| Molekylformel | C19 H15 N3 O8 S2 . 2 Na |
| Kvantitet | 25 mg |
| Synonym | Acid Red 35, 2,7-Naphthalenedisulfonic acid, 5-(acetylamino)-4-hydroxy-3-[2-(2-methylphenyl)diazenyl]-, sodium salt (1:2), 2,7-Naphthalenedisulfonic acid, 5-(acetylamino)-4-hydroxy-3-[(2-methylphenyl)azo]-, disodium salt (9CI), C.I. Acid Red 35 (7CI), C.I. Acid Red 35, disodium salt (8CI), Acid Brilliant Red 3B, Acid Fast Red 3B, Acid Fast Red 3B No. 855D, Acid Fast Red W 3B, Acid Magenta 6B, Acid Red 3B, Acid Red RR, Amacid Brilliant Red 5B, Atul Acid Red 3B, Belacid Red 3B, Best Acid Pink B, Brilliant Acid Red 3A, Brilliant Colacid Red 3A, Brilliant Fast Red S 3B, C.I. 18065, Colocid Brilliant Red 5B, Conacid Red CE, Diacid Supra Red 3B, Dinacid Fast Red 3B, Dycosacid Red 3B, Egacid Red R, Fast Red M 3B, Fenazo Light Red 3B, Kiton Fast Red B 2R, Midlon Red 3B, Multacid Red 3B, Optamine Red 3B, Peach Red 3B, Ritacid Red 3B, Suminol Fast Brilliant Red S 3B, Suminol Levelling Brilliant Red S 3B, Sumitomo Fast Red 3B, Supra Red 3B, Supracetil Red 3B, Supracid Red 3B, Supramin Red 3B, Supramine Red 3B, Supramine Red 3BA, Symulon Supramin Red 3B, Takaoka Acid Fast Red 3B, Tertracid Supra Red 3B, Triacid Fast Red 3BS, Vondacid Fast Red 3B, Vopsider Red ASTR 3A, Wool Red 3B |
| InChI formel | InChI=1S/C19H17N3O8S2.2Na/c1-10-5-3-4-6-14(10)21-22-18-16(32(28,29)30)8-12-7-13(31(25,26)27)9-15(20-11(2)23)17(12)19(18)24;;/h3-9,24H,1-2H3,(H,20,23)(H,25,26,27)(H,28,29,30);;/q;2*+1/p-2 |
| LEDER | [Na+].[Na+].CC(=O)Nc1cc(cc2cc(c(N=Nc3ccccc3C)c(O)c12)S(=O)(=O)[O-])S(=O)(=O)[O-] |
| IUPAC-namn | disodium;5-acetamido-4-hydroxy-3-[(E)-(2-methylphenyl)diazenyl]naphthalene-2,7-disulfonate |
| Molekylvikt (g/mol) | 523.447 |
| Formel vikt | 523.01 |