missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Violet 43 (Technical Grade), TRC
Specifications
Specifications
| Kemiskt namn eller material | Acid Violet 43 |
| CAS | 4430-18-6 |
| Molekylformel | C21 H14 N O6 S . Na |
| Kvantitet | 500 mg |
| Synonym | Benzenesulfonic acid, 2-[(9,10-dihydro-4-hydroxy-9,10-dioxo-1-anthracenyl)amino]-5-methyl-, monosodium salt (9CI), m-Toluenesulfonic acid, 6-[(4-hydroxy-1-anthraquinonyl)amino]-, monosodium salt (8CI), m-Toluenesulfonic acid, 6-[(4-hydroxy-1-anthraquinonyl)amino]-, sodium salt (7CI), 11362 Violet, 4-Hydroxy-1-(4-methyl-2-sulfoanilino)anthraquinone sodium salt, Acid Alizarine Violet R, Acid Violet 43, Acid Violet Anthraquinone, Ahcoquinone Blue IR, Alizarin Violanol R, Alizarine Brilliant Violet R, Alizarine Irisol R, Alizarine Irisol RD, Alizarine Light Violet B, Alizarine Light Violet RS, Alizarine Purple, Alizarine Purple RS, Alizarine Violanol R, Alizarine Violet 38017, Alizarine Violet NR, Allzurol Purple, Anthraquinone Iris R, C.I. 60730, C.I. Acid Violet 43, Chrome Violet Anthraquinone, Covasol Violet W 5002, Diacid Light Violet RS, Egacid Violet 3B, Elbenyl Violet C-RL, Erio Anthracene Cyanine JR, Erio Fast Cyanine JR, Erionyl Cyanine E-JR, Ext D&C Violet 2, Ext D and C Violet No. 2, Ext D&C Violet No. 2-38017, Ext D&C Violet No. 2-90131, Ext D&C Violet No. 2W098, External D and C Violet 2, External D&C Violet 2-308-139, External Violet 2, Fenalan Violet 3B, Hastings Light Fast Violet IRS, Japan Purple 401, Japan Purple No. 401, Japan Violet 401, Japan Violet No. 401, Kiton Fast Violet R, Purple No. 401, Solanol Violet 3B, Solway Purple, Solway Purple R, Solway Purple RA, Suminol Levelling Violet 3B, Supracen Violet 3B, Triacid Light Violet 3B, Unicert Violet K 7116J, Violet 401, Violet Anthraquinone, Violet No. 401, Vondacid Light Violet 3B |
| InChI formel | InChI=1S/C21H15NO6S.Na/c1-11-6-7-14(17(10-11)29(26,27)28)22-15-8-9-16(23)19-18(15)20(24)12-4-2-3-5-13(12)21(19)25;/h2-10,22-23H,1H3,(H,26,27,28);/q;+1/p-1 |
| LEDER | [Na+].Cc1ccc(Nc2ccc(O)c3C(=O)c4ccccc4C(=O)c23)c(c1)S(=O)(=O)[O-] |
| IUPAC-namn | sodium;2-[(4-hydroxy-9,10-dioxoanthracen-1-yl)amino]-5-methylbenzenesulfonate |
| Molekylvikt (g/mol) | 431.39 |
| Formel vikt | 431.044 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?