missing translation for 'onlineSavingsMsg'
Learn More

ACKR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11386-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16878423

  • 3313.85 SEK / 150µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a beta chemokine receptor, which is predicted to be a seven transmembrane protein similar to G protein-coupled receptors. Chemokines and their receptor-mediated signal transduction are critical for the recruitment of effector immune cells to the inflammation site. This gene is expressed in a range of tissues and hemopoietic cells. The expression of this receptor in lymphatic endothelial cells and overexpression in vascular tumors suggested its function in chemokine-driven recirculation of leukocytes and possible chemokine effects on the development and growth of vascular tumors. This receptor appears to bind the majority of beta-chemokine family members; however, its specific function remains unknown. This gene is mapped to chromosome 3p21.3, a region that includes a cluster of chemokine receptor genes.
Specifications

Specifications

ACKR2
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O00590, O08707, O09027
ACKR2, Atypical chemokine receptor 2, C C chemokine receptor D6, CCBP2, CCR10, CCR9, chemokine binding protein 2, Chemokine binding protein D6, Chemokine receptor CCR 10, Chemokine receptor CCR 9, CMKBR9, D6, hD6
Rabbit
IgG
150 μL
-20°C
Primary
1238, 140473, 59289
Immunohistochemistry (Paraffin)
Unconjugated
ACKR2
Liquid
Ackr2
ACKR2 Fusion Protein Ag1941
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers