Learn More
Micronova™ ACME Thread Adapters
Product Code. 12857033 Shop All Micronova Products
Change view
Click to view available options
Length (English):
5.75 in.
8 in.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
12857033 5.75 in. Each
12847033 8 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12857033 Supplier Micronova™ Supplier No. SSA3
  • ONLINE EXCLUSIVE PRICE
    636.00 SEK
    603.65 SEK / Paket med 1
    Save 32.35 SEK
    5% Av
 Restricted Item
Beräknad leverans: 01-04-2027
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy
The two versions of stainless steel ACME adapters connect threaded brush and mop adapters to Quick Disconnect, SSU Series Handles (SSA-3), or connect Quick Disconnect mop adapters to threaded handles (SSA-2).
The Micronova ACME Thread Adapters serve as the connection between various Micronova Adapters and either Threaded or Quick Disconnect handles. The SSA-2 serves to connect a range of Quick Disconnect mop adapters and cleaning tools with threaded handles, such as STA Series Handles. The SSA-3 serves to connect a range of threaded brushes and mop adapters, such as QPSL series, with the Quick Disconnect stainless steel SSU Series Handles.
  • ACME threaded adapters
  • Stainless steel with polyketone connectors
  • Connects various Adapters to either Threaded or Quick Disconnect handles
  • SSA-2 connects threaded handles to QD mop and tool adapters
  • SSA-3 connects threaded mop and brush adapters to QD SSU Handles
  • Autoclavable
Ordreinfo
Attach to a stainless-steel handle (sold separately) with 19-035-362B Thread Adapter.
Tekniske data
Product Type Male Thread Adapter
Length (English) 5.75 in.
Sterility Non-sterile
Length (Metric) 14.6 cm
Autoclavable Autoclavable
For Use With (Equipment) Threaded Connector for Handles and QD Mop Adapters and Tools
For Use With (Application) Floors, Walls, Isolators
Color-coded No
Material Electropolished Stainless Steel (304), Polyketone
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.