Läs mer
Acofide Trihydrate, TRC
Produktkod. 30325758
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30325758 250 mg
1 options
Produktkod. 30325758 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA190240250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Acofide Trihydrate
CAS 773092-05-0
Molekylformel C21H30N4O5S . (HCl) . 3(H2O)
Kvantitet 250 mg
Synonym Acotiamide Hydrochloride Trihydrate, N-(2-(Diisopropylamino)ethyl)-2-(2-hydroxy-4,5-dimethoxybenzamido)thiazole-4-carboxamide Hydrochloride Trihydrate.
LEDER O.O.O.Cl.COc1cc(O)c(cc1OC)C(=O)Nc2nc(cs2)C(=O)NCCN(C(C)C)C(C)C
Formel vikt 540.2021