missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Aconine, MedChemExpress
Beskrivning
Aconine inhibits receptor activator of nuclear factor (NF)-κB ligand (RANKL)-induced NF-κB activation.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Aconine |
| CAS | 509-20-6 |
| Färg | White |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C25H41NO9 |
| Kvantitet | 5 mg |
| Synonym | Jesaconine |
| Löslighetsinformation | DMSO : 100 mg/mL (200.16 mM; Need ultrasonic) |
| LEDER | O[C@@H]1C[C@H](OC)[C@@]23[C@]([H])(C4)[C@]5([H])[C@@H](O)[C@@]4(O)[C@@H](OC)[C@H](O)[C@]5(O)[C@H]6[C@H](OC)[C@@]([H])2[C@]1(COC)CN(CC)[C@H]63 |
| Molekylvikt (g/mol) | 499.59 |
| Visa mer |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?