missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOT7L Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | ACOT7L |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16886275
|
Proteintech
51016-2-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16876275
|
Proteintech
51016-2-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
ACOT7L | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
BACHL, ACOT7L, LOC344967 | |
LOC344967 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
344967 | |
Peptide | |
Primary | |
-20°C | |
LOC344967 |