missing translation for 'onlineSavingsMsg'
Learn More

ACOX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17571-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16827494

  • 3313.85 SEK / 150µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The product of this gene belongs to the acyl-CoA oxidase family. It encodes the branched-chain acyl-CoA oxidase which is involved in the degradation of long branched fatty acids and bile acid intermediates in peroxisomes. Deficiency of this enzyme results in the accumulation of branched fatty acids and bile acid intermediates, and may lead to Zellweger syndrome, severe mental retardation, and death in children.
Specifications

Specifications

ACOX2
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P97562, Q99424, Q9QXD1
ACOX2, BRCACOX, BRCOX, THCA CoA oxidase, THCCox
Rabbit
IgG
150 μL
-20°C
Primary
252898, 8309, 93732
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
ACOX2
Liquid
Acox2
ACOX2 Fusion Protein Ag11778
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers