Läs mer
MedChemExpress Acridine Orange 10-Nonyl Bromide
Specifikationer
Specifikationer
| Kvantitet | 10 mg |
| Förvaringskrav | 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light) |
| Löslighet | DMSO : 33.33 mg/mL (70.54 mM; Need ultrasonic) |
| Frakt skick | Room temperature |
| CAS | 75168-11-5 |
| Molekylvikt (g/mol) | 472.5 g/mol |
| Synonym | Nonylacridine orange |
| Mål | HBV |