Läs mer
Acridine Orange Stain, TRC
Produktkod. 30328451
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30328451 100 mg
1 options
Produktkod. 30328451 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA190933100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Acridine Orange Stain
CAS 494-38-2
Molekylformel C17 H19 N3
Kvantitet 100 mg
Synonym N3,N3,N6,N6-Tetramethyl-3,6-acridinediamine, 3,6-Bis(dimethylamino)acridine, 3,6-Di(dimethylamino)acridine, Acridine Orange Base, Acridine Orange Free Base, Brilliant Acridine Orange E, C.I. 46005B, C.I. Solvent Orange 15, NSC 194350, Solvent Orange 15, Waxoline Orange A
InChI formel InChI=1S/C17H19N3/c1-19(2)14-7-5-12-9-13-6-8-15(20(3)4)11-17(13)18-16(12)10-14/h5-11H,1-4H3
LEDER CN(C)c1ccc2cc3ccc(cc3nc2c1)N(C)C
IUPAC-namn 3-N,3-N,6-N,6-N-tetramethylacridine-3,6-diamine
Molekylvikt (g/mol) 265.36
Formel vikt 265.1579