Learn More
Cytiva (formerly Pall Lab) AcroPrep™ 96 Filter Plates, 1 mL
Optimized for a variety of applications requiring larger per well volume processing.
Brand: Cytiva (formerly Pall Lab) 5053
Additional Details : Weight : 0.65550kg
Description
- Plates are constructed of chemically resistant, biologically-inert polypropylene
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Glass Fiber (Borosilicate Glass Without Binder), Bio-Inert (Modified Low Protein-Binding Nylon) | |
Non-sterile | |
900 μL (Sample) |
3.0/0.2 μm | |
5/Pk. |