Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Nucleic Acid Binding
Optimized for a variety of applications requiring larger per well volume processing.
Brand:  Cytiva (formerly Pall Lab) 8133
Additional Details : Weight : 0.69350kg
Product Code. 17154621
  • 2350.00 SEK / Pack of 5
Estimated Shipment: 15-05-2024
to see stock.
Add to basket
Description
Description
  • Plates are constructed of chemically resistant, biologically-inert polypropylene
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
Silica Based Quartz Glass Fiber
Non-sterile
5/Pk.
Long Tips