Läs mer
Acryloyl-CoA, TRC
Produktkod. 30343192
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30343192 10 mg
1 options
Produktkod. 30343192 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA67758010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Acryloyl-CoA
CAS 5776-58-9
Molekylformel C24 H38 N7 O17 P3 S
Kvantitet 10 mg
Synonym Acryloyl Coenzyme A, Acryloyl-CoA, Acrylyl Coenzyme A, Thioacrylic Acid S-ester With Coenzyme A
InChI formel InChI=1S/C24H38N7O17P3S/c1-4-15(33)52-8-7-26-14(32)5-6-27-22(36)19(35)24(2,3)10-45-51(42,43)48-50(40,41)44-9-13-18(47-49(37,38)39)17(34)23(46-13)31-12-30-16-20(25)28-11-29-21(16)31/h4,11-13,17-19,23,34-35H,1,5-10H2,2-3H3,(H,26,32)(H,27,36)(H,40,41)(H,42,43)(H2,25,28,29)(H2,37,38,39)/t13-,17-,18-,19-,23-/m1/s1
LEDER CC(C)(COP(=O)(O)OP(=O)(O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1OP(=O)(O)O)n2cnc3c(N)ncnc23)[C@H](O)C(=O)NCCC(=O)NCCSC(=O)C=C
IUPAC-namn S-[2-[3-[[(2S)-4-[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-3-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl]oxy-2-hydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyl] prop-2-enethioate
Molekylvikt (g/mol) 821.582
Formel vikt 821.126