Learn More
Acryloylglycine, TRC
Specifications
Specifications
| Kemiskt namn eller material | Acryloylglycine |
| CAS | 24599-25-5 |
| Molekylformel | C5 H7 N O3 |
| Kvantitet | 250 mg |
| Synonym | N-(1-Oxo-2-propen-1-yl)glycine, N-(1-Oxo-2-propenyl)glycine, Acryloylaminoacetic Acid, N-(Carboxymethyl)acrylamide, N-Acryloylglycine, NSC 288735 |
| InChI formel | InChI=1S/C5H7NO3/c1-2-4(7)6-3-5(8)9/h2H,1,3H2,(H,6,7)(H,8,9) |
| LEDER | OC(=O)CNC(=O)C=C |
| IUPAC-namn | 2-(prop-2-enoylamino)acetic acid |
| Molekylvikt (g/mol) | 129.11 |
| Formel vikt | 129.0426 |