missing translation for 'onlineSavingsMsg'
Learn More

ACTC1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1F2B9, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66125-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16877715

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is specific to the ACTC1. It does not cross-react with other actin isoforms.

ACTC1 are part of actins, which are highly conserved proteins that are involved in various types of cell motility. Polymerization of globular actin (G-actin) leads to a structural filament (F-actin) in the form of a two-stranded helix. Each actin can bind to four others. The protein encoded by this gene belongs to the actin family which is comprised of three main groups of actin isoforms, alpha, beta, and gamma. The alpha actins are found in muscle tissues and are a major constituent of the contractile apparatus. Defects in this gene have been associated with idiopathic dilated cardiomyopathy (IDC) and familial hypertrophic cardiomyopathy (FHC).
Specifications

Specifications

ACTC1
1F2B9
Unconjugated
ACTC1
Liquid
ACTC1
Peptide
Protein G
RUO
Antibody
Monoclonal
Human, Porcine, Mouse, Rat
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)
0.75 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P68032, P68033, P68035
ACTC, ACTC1, ACTC1 cardiac muscle 1, ACTC1 specific, Actin cardiac muscle specific, Actin, alpha cardiac muscle 1, actin, alpha, cardiac muscle 1, Alpha cardiac actin, CMD1R, CMH11
Mouse
IgG1
20 μL
-20°C
Primary
100152267, 11464, 29275, 70
Videos
SDS
Documents
Certificates
Special Offers

Special Offers