missing translation for 'onlineSavingsMsg'
Learn More

Activated Charcoal, Acid-washed with Hydrochloric Acid, Honeywell Fluka™

Catalog Number p-7120118
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Drum with liner
Poly bottle
This item is not returnable. View return policy
7440-44-0
C
12.011
MFCD00133992
OKTJSMMVPCPJKN-UHFFFAOYSA-N
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
5462310
CHEBI:27594
carbon
[C]
This item is not returnable. View return policy
7440-44-0
C
12.011
MFCD00133992
OKTJSMMVPCPJKN-UHFFFAOYSA-N
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
5462310
CHEBI:27594
carbon
[C]

Acid-washed with hydrochloric acid

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Color Black
pH 6-9
Odor Odorless
Packaging Poly bottle
Quantity 500 g
UN Number NONH for all modes of transport
Formula Weight 12.01g/mol
Physical Form Powder
Chemical Name or Material Activated charcoal, Acid-washed with hydrochloric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'