missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activated Charcoal, Norit™, SX ultra, U.S. Food chemicals codex, Honeywell Fluka™
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic Bottle
Description
- Norit is a registered trademark of Norit N.V.
Specifications
Color | Black |
pH | 6 to 9 |
Odor | Odorless |
Packaging | Plastic Bottle |
Quantity | 250 g |
UN Number | NONH for all modes of transport |
Beilstein | 4360473 |
Formula Weight | 12.01g/mol |
Chemical Name or Material | Activated Charcoal Norit™, Norit SX ultra |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'