missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACVR2B Human Protein
ACVR2B is a type II member of the TGF-beta family of receptor ser/thr kinases.
Brand: Invitrogen™ PR9140B
Additional Details : Weight : 0.01000kg
Description
ACVR2B is involved in the activin and myostatin signaling pathway.
Specifications
NP_001097.2 | |
62kDa | |
Kinase proteins are shipped on dry ice and should be stored at -80°C. | |
Catalytic | |
STKR Kinase Family | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
100μg | |
RUO - research use only | |
ACVR2B | |
Human | |
Baculovirus |