Learn More
Invitrogen™ ACVR2B Human Protein
ACVR2B is a type II member of the TGF-beta family of receptor ser/thr kinases.
5340.00 SEK - 79000.00 SEK
Specifications
Accession Number | NP_001097.2 |
---|---|
Molecular Weight (g/mol) | 62kDa |
Storage Requirements | Kinase proteins are shipped on dry ice and should be stored at -80°C. |
Structural Form | Catalytic |
Gene Alias | ACVR2B |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10477318
|
Invitrogen™
PV6049 |
10μg |
5340.00 SEK
Each |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15498713
|
Invitrogen™
PR9140B |
100μg |
18260.00 SEK
100µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15478803
|
Invitrogen™
PV6050 |
1mg |
79000.00 SEK
1mg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ACVR2B is involved in the activin and myostatin signaling pathway.
Specifications
NP_001097.2 | |
Kinase proteins are shipped on dry ice and should be stored at -80°C. | |
ACVR2B | |
Human | |
Baculovirus |
62kDa | |
Catalytic | |
STKR Kinase Family | |
GST Tag |