Learn More
Invitrogen™ Human ACVRL1 (ALK1), GST Tag Recombinant Protein
Human ACVRL1 (ALK1), GST Tag Recombinant Protein
Brand: Invitrogen™ PR8914A
Additional Details : Weight : 0.01000kg
Description
ACVRL1, similar to other type 1 receptors, has a conserved serine-threonine kinase subdomain, a glycine- and serine-rich region (called the GS domain) preceding the kinase domain, and a short C-terminal tail. Mutations in ACVRL1 are associated with hemorrhagic telangiectasia type 2.
Specifications
NP_000011.2 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Polysorbate 20, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
ACVRL1 (ALK1) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
68.9kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
STKR Kinase Family | |
Recombinant | |
Baculovirus |