Learn More
Abnova™ ACY1 (Human) Recombinant Protein
Human ACY1 full-length ORF ( AAH00545, 1 a.a. - 408 a.a.) recombinant protein with GST-tag at N-terminal.
Brand:  Abnova™ H00000095-P01.25ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16131251
  • 6845.00 SEK / 25µg
Estimated Shipment: 28-06-2024
to see stock.
Description
Description
  • Sequence: MTSKGPAEEHPSVTLFRQYLRIRTVQPKPDYGAAVAFFEETARQLGLGCQKVEVAPGYVVTVLTWPGTNPTLSSILLNSHTDVVPVFKEHWSHDPFEAFKDSEGYIYARGAQDVKCVSIQYLEAVRRLKVEGHRFPRTIHMTFVPDEEVGGHQGMELFVQRPEFHALRAGFALDEGIANPTDAFTVFYSERSPWWVRVTSTGRPGHASRFMEDTAAEKLHKVVNSILAFREKEWQRLQSNPHLKEGSVTSVNLTKLEGGVAYNVIPATMSASFDFRVAPDVDFKAFEEQLQSWCQAAGEGVTLEFAQKWMHPQVTPTDDSNPWWAAFSRVCKDMNLTLEPEIMPAATDNRYIRAVGVPALGFSPMNRTPVLLHDHDERLHEAVFLRGVDIYTRLLPALASVPALPSDS
Specifications
Specifications
AAH00545
aminoacylase 1
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACY1
GST
95
Wheat germ expression system
25 μg
ACY1D, ACYLASE
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer