Läs mer
Acyclovir Impurity O (N-Hydroxymethyl Acyclovir), Technical Grade, TRC
Produktkod. 30137608
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137608 100 mg
30136021 50 mg
17912755 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137608 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA194335100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2190509-40-9
Molekylformel C9 H13 N5 O4
Molekylvikt (g/mol) 255.231
Synonym 9-[(2-Hydroxyethoxy)-methyl]-2-[(hydroxymethyl)amino]-1,9-dihydro-6H-purin-6-one
IUPAC-namn 9-(2-hydroxyethoxymethyl)-2-(hydroxymethylamino)-1H-purin-6-one
LEDER OCCOCn1cnc2C(=O)NC(=Nc12)NCO
Specifikationer
Kemiskt namn eller material Acyclovir Impurity O (N-Hydroxymethyl Acyclovir), Technical Grade
Analyt- eller komponentnamn Acyclovir Impurity O (N-Hydroxymethyl Acyclovir), Technical Grade
Frakt skick Room Temperature
InChI formel InChI=1S/C9H13N5O4/c15-1-2-18-5-14-3-10-6-7(14)12-9(11-4-16)13-8(6)17/h3,15-16H,1-2,4-5H2,(H2,11,12,13,17)
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 100 mg
Formel vikt 255.097