missing translation for 'onlineSavingsMsg'
Learn More

ADAMTSL4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15304-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824434

  • 3248.87 SEK / 150µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a member of ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs)-like gene family and encodes a protein with protein with seven thrombospondin type 1 repeats. The thrombospondin type 1 repeat domain is found in many proteins with diverse biological functions including cellular adhesion, angiogenesis, and patterning of the developing nervous system. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications

Specifications

ADAMTSL4
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q4FZU4, Q6UY14, Q80T21
ADAMTS like 4, ADAMTS like protein 4, ADAMTSL 4, ADAMTSL4, TSRC1, UNQ2803/PRO34012
Rabbit
IgG
150 μL
-20°C
Primary
229595, 310670, 54507
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
ADAMTSL4
Liquid
Adamtsl4
ADAMTSL4 Fusion Protein Ag7781
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers