Learn More
HUBER Adapter for Bath
Accessory for thermostats.
Brand: HUBER 6359
Additional Details : Weight : 0.05000kg
Description
- Adapter
Specifications
Thread Size: M16 x 1 female to M16 x 1 female | |
Thermostats | |
Adapter |