Learn More
HUBER Adapter for Bath
Accessory for thermostats.
Brand:  HUBER 6442
Additional Details : Weight : 0.18000kg
 View more versions of this product
Product Code. 10289741
  • 998.52 SEK / Each
Estimated Shipment: 28-06-2024
to see stock.
Add to basket
Description
Description
  • Adapter
Specifications
Specifications
Thread Size: M30 x 1.5 male to G3/4 female
Adapter