Learn More
HUBER Adapter for Bath
Accessory for thermostats.
Brand:  HUBER 6444
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 10299741
  • 1197.80 SEK / Each
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
Description
Description
  • Adapter
Specifications
Specifications
Thread Size: M30 x 1.5 male to G 1 male
Adapter
16 x 1 m