Learn More
HUBER Adapter for Bath
Accessory for thermostats.
Brand:  HUBER 9243
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10783276
  • 1250.80 SEK / Each
Estimated Shipment: 28-06-2024
to see stock.
Add to basket
Description
Description
  • Adapter
Specifications
Specifications
Thread Size: M24 x 1.5 male to RP 1/2 female
Adapter
16 x 1 m