Learn More
Hettich™ Adapter for Carrier 5051 with Decanting Aid
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
POA
Specifications
Type | Adapter with Decanting Aid |
---|---|
Max. Tube Length | 100 mm |
For Use With (Equipment) | Carrier 5051 |
Description
- Adapter for Carrier 5051
- With Decanting Aid
- For use with Hettich Rotors
- Compatible with a wide range of tubes to fit a variety of applications
Specifications
Adapter with Decanting Aid | |
100 mm | |
Carrier 5051 |