missing translation for 'onlineSavingsMsg'
Learn More

Hettich™ Adapter for Carrier no. 1427

Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life

Brand:  Hettich™ 5230

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 12003924

  • 1030.00 SEK / Each
Estimated Shipment: 13-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Adapter for Carrier no. 1427
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications

Specifications

Rotor Adapter
12 x 4.5 to 7 mL Tubes
100 mm
12
Blood/Urine, Round Bottom
12 mm
Carrier no. 1427
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers