Learn More
Hettich™ Adapter for Carrier no. 5092
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 5122
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10563903
  • 1515.00 SEK / Each
Estimated Shipment: 11-06-2024
to see stock.
Description
Description
  • Adapter for Carrier no. 5092
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
Rotor Adapter
4 x 30 mL
110 mm
4
Round Bottom, Conical, Skirted
25 mm
Carrier no. 5092