Learn More
Hettich™ Adapter for Carrier no. 5092
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 5120
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11485098
  • 1095.00 SEK / Each
Estimated Shipment: 28-06-2024
to see stock.
Description
Description
  • Adapter for Carrier no. 5092
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
Rotor Adapter
12 x 4 to 7 mL Tubes
100 mm
12
Round Bottom, Conical, Blood/Urine
13 mm
Carrier no. 5092