Learn More
Hettich™ Adapter for Carrier no. 5092
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 5135
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 12083914
  • 1175.00 SEK / Each
Estimated Shipment: 01-07-2024
to see stock.
Description
Description
  • Adapter for Carrier no. 5092
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
Rotor Adapter
2 x 50 mL
115 mm
2
Skirted Conical
29 mm
Carrier no. 5092