Learn More
Hettich™ Adapter for Carrier no. 5092
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 5136
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12356232
  • 1515.00 SEK / Each
Estimated Shipment: 01-07-2024
to see stock.
Description
Description
  • Adapter for Carrier no. 5092
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
Rotor Adapter
8 x 4 to 10 mL Tubes
102 mm
8
Blood/Urine, Round Bottom
17 mm
Carrier no. 5092