Learn More
Hettich™ Adapter for Carrier no. 5092
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand: Hettich™ 5134
Additional Details : Weight : 0.10000kg
Description
- Adapter for Carrier no. 5092
- For use with Hettich Rotors
- Compatible with a wide range of tubes to fit a variety of applications
Specifications
Rotor Adapter | |
3 x 25 mL | |
90 mm | |
3 |
Skirted Conical | |
25 mm | |
Carrier no. 5092 |