Learn More
Benchmark Scientific Adapter for Cryovials/HPLC Vials
Compatible with myFuge™ 5 Mini Centrifuge
Brand: Benchmark Scientific C1005-AC2
Additional Details : Weight : 0.50000kg
Description
- Adapter pack for myFuge™ 5 Mini Centrifuge
- Holds 0.5-2 mL Cryovials and 1.5/2.0 mL HPLC vials
- Includes 4 Adapters per pack
Specifications
1/Adapter | |
Non-sterile | |
4 |
Neoprene Rubber | |
No | |
No |